(±)-3-(Boc-amino)piperidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:172603-05-3
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD03839941
:200.28 g/mol
Melting Pt:102…108 °C
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09
Storage Temperature:Refrigerator