Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3-methoxypyrazine
2-Chloro-3-methoxypyrazine
2-Chloro-3-methoxypyrazine
2-Chloro-3-methoxypyrazine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-3-methoxypyrazine
Supplier:
Apollo Scientific
About this item
CAS Number:
40155-28-0
:
Formula:
C₅H₅ClN₂O
MDL Number:
MFCD02684324
:
144.5600 g/mol
Melting Pt:
31…32 °C
:
GHS07
Storage Temperature:
Refrigerator