Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Tyrosine methyl ester hydrochloride
L-Tyrosine methyl ester hydrochloride 98%
L-Tyrosine methyl ester hydrochloride 98%
:
Apollo Scientific
L-Tyrosine methyl ester hydrochloride 98%
:
Apollo Scientific
:
3417-91-2
:
:
C₁₀H₁₃NO₃
:
MFCD00012607
:
231.68 g/mol
:
190…193 °C (dec.)
:
GHS07
:
Refrigerator