
4-Ethynylpyridine
Catalog # APOSOR60207-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:103.12 g/mol
Melting Pt:98 °C
:GHS07
Storage Temperature:Freezer
CAS Number:2510-22-7
Formula:C₇H₅N
MDL Number:MFCD04038420