Order Entry
Germany
ContactUsLinkComponent
 
Irritant
1-Phenylcyclopropanamine hydrochloride 95%
Catalog # APOSOR60203-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Phenylcyclopropanamine hydrochloride 95%
Catalog # APOSOR60203-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :169.6500 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:73930-39-9
Formula:C₉H₁₂ClN
MDL Number:MFCD07995722