Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Borono-DL-phenylalanine
4-Borono-DL-phenylalanine
4-Borono-DL-phenylalanine
Irritant
4-Borono-DL-phenylalanine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Borono-DL-phenylalanine
Supplier:
Apollo Scientific
About this item
CAS Number:
90580-64-6
:
Formula:
C₉H₁₂BNO₄
MDL Number:
MFCD01863704
:
209.01 g/mol
Melting Pt:
270 °C (dec.)
:
GHS07
Storage Temperature:
Refrigerator