3-(2-Bromophenyl)-5-(3-nitrophenyl)-1,2,4-oxadiazole
Catalog # APOSOR59386-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:1000339-27-4
Formula:C₁₄H₈BrN₃O₃
MDL Number:MFCD09878377