
Dimethylcyclobutane-1,1-dicarboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10224-72-3
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00082719
:172.180 g/mol
Boiling Pt:122 °C/5mmHg
Density:1.118 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient