Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-methoxypyridine
3-Bromo-5-methoxypyridine 98%
3-Bromo-5-methoxypyridine 98%
3-Bromo-5-methoxypyridine 98%
APOSOR5842-100G
:
Apollo Scientific
:
3-Bromo-5-methoxypyridine 98%
APOSOR5842-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
188.02 g/mol
:
29…35 °C
:
102 °C
:
GHS05
:
GHS07
:
Ambient
:
50720-12-2
:
C₆H₆BrNO
:
MFCD00234169