Order Entry
Germany
ContactUsLinkComponent
Methyl thiazole-4-carboxylate 97%
Irritant
Methyl thiazole-4-carboxylate 97%
Catalog # APOSOR5602-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl thiazole-4-carboxylate 97%
Catalog # APOSOR5602-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.1600 g/mol
Melting Pt:38…42 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:59418-09-6
Formula:C₅H₅NO₂S
MDL Number:MFCD06410940