Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl mercaptan sodium salt
Methyl mercaptan sodium salt solution
Methyl mercaptan sodium salt solution
Flammable
Corrosive
Methyl mercaptan sodium salt solution
Supplier:
Apollo Scientific
Certificates
Expand image
Flammable
Corrosive
Methyl mercaptan sodium salt solution
Supplier:
Apollo Scientific
About this item
CAS Number:
5188-07-8
:
Formula:
CH₃NaS
MDL Number:
MFCD00174316
:
70.08 g/mol
UN:
2920
ADR:
8+3,I
:
GHS02
:
GHS05
Storage Temperature:
Ambient