Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9-Aminofluorene hydrochloride
9-Aminofluorene hydrochloride
9-Aminofluorene hydrochloride
Irritant
9-Aminofluorene hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
9-Aminofluorene hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
5978-75-6
:
Formula:
C₁₃H₁₁N
MDL Number:
MFCD00012536
:
217.70 g/mol
Melting Pt:
265…370 °C (dec.)
:
GHS07
Storage Temperature:
Ambient