Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Irritant
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
35969-51-8
:
Formula:
C₁₀H₁₁NO₄
MDL Number:
MFCD00663817
:
209.2 g/mol
:
GHS07
Storage Temperature:
Ambient