
4-Hydroxy-6-methyl-2-pyrone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:675-10-5
:
Formula:C₆H₆O₃
MDL Number:MFCD00006641
:126.11 g/mol
Melting Pt:188…190 °C (dec.)
EINECS:211-619-2
:GHS07
Storage Temperature:Ambient