Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Auftragserfassung
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(tert-Butoxycarbonyl)-1,5-diaminopentan
N-(tert-Butoxycarbonyl)-1,5-diaminopentan 98%
N-(tert-Butoxycarbonyl)-1,5-diaminopentan 98%
:
Apollo Scientific
N-(tert-Butoxycarbonyl)-1,5-diaminopentan 98%
:
Apollo Scientific
:
51644-96-3
:
:
C₁₀H₂₂N₂O₂
:
MFCD00210020
:
202.30 g/mol
:
0,972 g/cm³ (25 °C)
:
109 °C
:
Raumtemperatur
:
2735
:
8,III
:
GHS05
:
GHS07