
Methyl-3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Catalog # APOSOR52105-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.07 g/mol
Melting Pt:156…157 °C
:GHS07
Storage Temperature:Ambient
CAS Number:342613-63-2
Formula:C₉H₇BrN₂O₂
MDL Number:MFCD10566741