
6-(Tetrahydro-2H-pyran-4-yloxy)pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:906352-78-1
:
Formula:C₁₁H₁₃NO₄
MDL Number:MFCD11100042
:223.23 g/mol
Melting Pt:111…112.5 °C
:GHS07
Storage Temperature:Ambient