Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Methyl-2-pyridinyl)methanamine
(3-Methyl-2-pyridinyl)methanamine
(3-Methyl-2-pyridinyl)methanamine
(3-Methyl-2-pyridinyl)methanamine
Supplier:
Apollo Scientific
Certificates
Expand image
(3-Methyl-2-pyridinyl)methanamine
Supplier:
Apollo Scientific
About this item
CAS Number:
153936-26-6
:
Formula:
C₇H₁₀N₂
MDL Number:
MFCD04116209
ADR:
8,III
:
122.17 g/mol
UN:
2735
:
GHS05
:
GHS07
Storage Temperature:
Ambient