Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Hydroxy-1-naphthoic acid
6-Hydroxy-1-naphthoic acid
6-Hydroxy-1-naphthoic acid
Irritant
6-Hydroxy-1-naphthoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Hydroxy-1-naphthoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
2437-17-4
:
Formula:
C₁₁H₈O₃
MDL Number:
MFCD00209983
:
188.1800 g/mol
Melting Pt:
210 °C
:
GHS07
Storage Temperature:
Refrigerator