2'-Methoxyacetophenone 98%
New Product
: 1-(2-Methoxyphenyl)ethanone
- :579-74-8
- :C₉H₁₀O₂
- :MFCD00008725
- :150.18 g/mol
- :131 °C/18mmHg
- :1.09 g/cm³ (25 °C)
- :108 °C
- :Ambient
- :209-446-2
: 1-(2-Methoxyphenyl)ethanone
- :579-74-8
- :C₉H₁₀O₂
- :MFCD00008725
- :150.18 g/mol
- :131 °C/18mmHg
- :1.09 g/cm³ (25 °C)
- :108 °C
- :Ambient
- :209-446-2
Product Family Options
- Pack typePk
- New ProductNew Product
- New ProductNew Product
- New ProductNew Product
- New ProductNew Product