
2-Formyl-6-methoxypyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54221-96-4
:
Formula:C₇H₇NO₂
MDL Number:MFCD05664030
:137.14 g/mol
Boiling Pt:103…104 °C/20mmHg
Density:1.14 g/cm³ (25 °C)
Flash Pt:81 °C
:GHS07
Storage Temperature:Ambient