Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloronicotinic acid
6-Chloronicotinic acid 98%
6-Chloronicotinic acid 98%
6-Chloronicotinic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloronicotinic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
5326-23-8
:
Formula:
C₆H₄ClNO₂
MDL Number:
MFCD00006241
:
157.56 g/mol
Melting Pt:
190…192 °C (dec.)
EINECS:
226-201-5
:
GHS07
Storage Temperature:
Ambient