2-Ethoxynicotinic acid 98%
Catalog # APOSOR5067-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:167.16 g/mol
Melting Pt:90…92 °C
:GHS07
Storage Temperature:Ambient
CAS Number:35969-54-1
Formula:C₈H₉NO₃
MDL Number:MFCD00128788