N-Benzylidene-tert-butylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6852-58-0
:
Formula:C₁₁H₁₅N
MDL Number:MFCD00043553
:161.24 g/mol
Boiling Pt:209 °C
Melting Pt:–30 °C
Density:0.906 g/cm³ (25 °C)
Flash Pt:78 °C
:GHS07
:GHS08
Storage Temperature:Ambient