
Methyl-3-aminopyrazine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16298-03-6
:
Formula:C₆H₇N₃O₂
MDL Number:MFCD00010097
:153.14 g/mol
Melting Pt:166…172 °C
EINECS:240-387-5
:GHS07
Storage Temperature:Ambient