
tert-Butyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916420-27-4
:
Formula:C₁₂H₁₅Cl₂N₃O₂
MDL Number:MFCD09025732
:304.1700 g/mol
Melting Pt:79…81 °C
:GHS07
Storage Temperature:Ambient