Order Entry
Germany
ContactUsLinkComponent
2,4-Diaminopyrimidine-5-carbonitrile 95%
2,4-Diaminopyrimidine-5-carbonitrile 95%
Catalog # APOSOR480761-5G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Diaminopyrimidine-5-carbonitrile 95%
Catalog # APOSOR480761-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :135.13 g/mol
Storage Temperature:Ambient
CAS Number:16462-27-4
Formula:C₅H₅N₅
MDL Number:MFCD00085600