2-(2-Bromphenyl)ethanol 98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
- Boiling Pt:97 °C/0,675mmHg
- Density:1,483 g/cm³ (25 °C)
- Flash Pt:>113 °C
- Storage Temperature:Raumtemperatur
6 Options Available Below
Product Details & Documents
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
- Boiling Pt:97 °C/0,675mmHg
- Density:1,483 g/cm³ (25 °C)
- Flash Pt:>113 °C
- Storage Temperature:Raumtemperatur
Product Family Options
Product Information
- PackungVEAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



