Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Isoquinoline-6-carbonitrile
Isoquinoline-6-carbonitrile
Isoquinoline-6-carbonitrile
Isoquinoline-6-carbonitrile
APOSOR48060-250MG
:
Apollo Scientific
:
Isoquinoline-6-carbonitrile
APOSOR48060-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
154.1680 g/mol
:
GHS07
:
Refrigerator
:
106778-42-1
:
C₁₀H₆N₂
:
MFCD09991660