
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:258278-25-0
:
Formula:C₂₇H₃₉ClN₂
MDL Number:MFCD07369796
:427.07 g/mol
Melting Pt:289…293 °C
:GHS07
Storage Temperature:Ambient