Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1H-Pyrazol-1-ylmethyl)aniline
4-(1H-Pyrazol-1-ylmethyl)aniline
4-(1H-Pyrazol-1-ylmethyl)aniline
APOSOR470965-5G
:
Apollo Scientific
:
4-(1H-Pyrazol-1-ylmethyl)aniline
APOSOR470965-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
173.22 g/mol
:
Ambient
:
142335-61-3
:
C₁₀H₁₁N₃
:
MFCD03422514