
1,5-Dimethyl-1H-pyrazol-3-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35100-92-6
:
Formula:C₅H₉N₃
MDL Number:MFCD04970041
Boiling Pt:98 °C/3mmHg
Melting Pt:65…68 °C
:111.15 g/mol
:GHS07
Storage Temperature:Ambient