Order Entry
Germany
ContactUsLinkComponent
2,4-Dichloropyrimidine-5-carboxylic acid amide 95%
2,4-Dichloropyrimidine-5-carboxylic acid amide 95%
Catalog # APOSOR46232-5G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloropyrimidine-5-carboxylic acid amide 95%
Catalog # APOSOR46232-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :192.0028 g/mol
Melting Pt:195 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:1240390-28-6
Formula:C₅H₃Cl₂N₃O
MDL Number:MFCD16658133