
2-(Boc-amino)pyridine-5-carboxaldehyde
Catalog # APOSOR46112-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.25 g/mol
Melting Pt:174…176 °C
:GHS07
Storage Temperature:Ambient
CAS Number:199296-40-7
Formula:C₁₁H₁₄N₂O₃
MDL Number:MFCD08064228