
Methyl (S)-piperidine-2-carboxylate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18650-39-0
:
Formula:C₇H₁₄ClNO₂
MDL Number:MFCD00672366
:179.6446 g/mol
Melting Pt:176…177 °C
:GHS07
Storage Temperature:Ambient