
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride 98%
Catalog # APOSOR460007-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:179.644 g/mol
Melting Pt:159…165 °C
:GHS07
Storage Temperature:Ambient
CAS Number:18650-38-9
Formula:C₇H₁₄ClNO₂
MDL Number:MFCD00237412