Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-1-Boc-3-hydroxypiperidine
(R)-1-Boc-3-Hydroxypiperidine 98%
(R)-1-Boc-3-Hydroxypiperidine 98%
:
Apollo Scientific
(R)-1-Boc-3-Hydroxypiperidine 98%
:
Apollo Scientific
:
143900-43-0
:
:
C₁₀H₁₉NO₃
:
MFCD04115306
:
201.27 g/mol
:
43…50 °C
:
104 °C
:
Ambient
:
2811
:
6.1,III
:
GHS06