tert-Butyl-4-hydroxyazepane-1-carboxylate
Catalog # APOSOR451215-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:215.2900 g/mol
Storage Temperature:Ambient
CAS Number:478832-21-2
Formula:C₁₁H₂₁NO₃
MDL Number:MFCD07368610