Order Entry
Germany
ContactUsLinkComponent
 
(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate
Catalog # APOSOR451105-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate
Catalog # APOSOR451105-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.35 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:859518-32-4
Formula:C₁₅H₂₂N₂O₂
MDL Number:MFCD04972241