Order Entry
Germany
ContactUsLinkComponent
7-Bromoindazole-1-carboxylic acid tert-butyl ester 98%
7-Bromoindazole-1-carboxylic acid tert-butyl ester 98%
Catalog # APOSOR43565-250MG
Supplier:  Apollo Scientific
CAS Number:  
7-Bromoindazole-1-carboxylic acid tert-butyl ester 98%
Catalog # APOSOR43565-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :297.16 g/mol
Storage Temperature:Ambient
CAS Number:1092352-37-8
Formula:C₁₂H₁₃BrN₂O₂
MDL Number:MFCD11505878