Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Irritant
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-tert-Butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
862728-60-7
:
Formula:
C₉H₁₃N₅
MDL Number:
MFCD00128294
:
191.24 g/mol
:
GHS07
Storage Temperature:
Ambient