1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1003846-21-6
:
Formula:C₁₄H₂₃BN₂O₃
MDL Number:MFCD16556148
:278.1550 g/mol
:GHS07
Storage Temperature:Refrigerator