
4-Bromo-o-anisaldehyde 98%
Catalog # APOSOR40709-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:215.05 g/mol
Melting Pt:67…71 °C
:GHS07
Storage Temperature:Ambient
CAS Number:43192-33-2
Formula:C₈H₇BrO₂
MDL Number:MFCD08276823