
Orotic acid monohydrate
Catalog # APOSOR40619-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:156.10 g/mol (anhy)
Melting Pt:334 °C
EINECS:200-619-8
:GHS07
Storage Temperature:Ambient
CAS Number:50887-69-9
Formula:C₅H₄N₂O₄·1H₂O
MDL Number:MFCD00149398