Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate 95+%
Irritant
tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate 95+%
Catalog # APOSOR40315-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate 95+%
Catalog # APOSOR40315-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :225.29 g/mol
Melting Pt:91…92 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:331281-25-5
Formula:C₁₁H₁₉N₃O₂
MDL Number:MFCD08234564