Order Entry
Germany
ContactUsLinkComponent
Isoquinoline-3-carbaldehyde 95%
Irritant
Isoquinoline-3-carbaldehyde 95%
Catalog # APOSOR40275-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Isoquinoline-3-carbaldehyde 95%
Catalog # APOSOR40275-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :157.1700 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:5470-80-4
Formula:C₁₀H₇NO
MDL Number:MFCD06227437