Order Entry
Germany
ContactUsLinkComponent
Dimethylcyclopropane-1,1-dicarboxylate 99%
Dimethylcyclopropane-1,1-dicarboxylate 99%
Catalog # APOSOR40111-1G
Supplier:  Apollo Scientific
CAS Number:  
Dimethylcyclopropane-1,1-dicarboxylate 99%
Catalog # APOSOR40111-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :158.15 g/mol
Boiling Pt:196…198 °C
Density:1.147 g/cm³ (25 °C)
Flash Pt:95 °C
EINECS:414-240-2
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:6914-71-2
Formula:C₇H₁₀O₄
MDL Number:MFCD00192079