4-Chloro-3-nitrobenzyl alcohol
Catalog # APOSOR401027-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:187.58 g/mol
Storage Temperature:Ambient
CAS Number:55912-20-4
Formula:C₇H₆ClNO₃
MDL Number:MFCD00007086