Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-methoxypyrazine
2-Amino-5-methoxypyrazine 98%
2-Amino-5-methoxypyrazine 98%
2-Amino-5-methoxypyrazine 98%
APOSOR40076-1G
:
Apollo Scientific
:
2-Amino-5-methoxypyrazine 98%
APOSOR40076-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
125.13 g/mol
:
Ambient
:
54013-07-9
:
C₅H₇N₃O
:
MFCD09909647