Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxy-4-nitrobenzaldehyde
3-Methoxy-4-nitrobenzaldehyde 97%
3-Methoxy-4-nitrobenzaldehyde 97%
Irritant
3-Methoxy-4-nitrobenzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Methoxy-4-nitrobenzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
80410-57-7
:
Formula:
C₈H₇NO₄
MDL Number:
MFCD08703367
:
181.1455 g/mol
:
GHS07
Storage Temperature:
Ambient